Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylamino-1-propanol, 99%

Catalog No. p-115580
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

CAS: 3179-63-3 | C5H13NO | 103.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3179-63-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00002945
InChI Key PYSGFFTXMUWEOT-UHFFFAOYSA-N
Synonym 3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol
PubChem CID 76646
IUPAC Name 3-(dimethylamino)propan-1-ol
SMILES CN(C)CCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless to Yellow
Density 0.8860g/mL
Boiling Point 159.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.4370
Linear Formula (CH3)2N(CH2)3OH
Quantity 100 g
Beilstein 04, I, 433
Specific Gravity 0.886
Solubility Information Solubility in water: soluble
Viscosity 6.85 mPa.s (20°C)
Formula Weight 103.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Dimethylamino-1-propanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 221-659-2
RTECSNumber UB3155000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.