Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193814
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 3-(Dimethylamino)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride)
CAS 1256355-23-3
Color White
Molecular Formula C8H13BClNO2
Quantity 1 g
MDL Number MFCD04112542
Synonym 3-(Dimethylamino)benzeneboronic Acid Hydrochloride
InChI Key QDYZZIRCCVRLBI-UHFFFAOYSA-N
SMILES Cl.CN(C)C1=CC=CC(=C1)B(O)O
IUPAC Name [3-(dimethylamino)phenyl]boronic acid hydrochloride
Molecular Weight (g/mol) 201.46
PubChem CID 44754897
Formula Weight 201.46
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.