Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Dimethylaminophenol, 97+%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents, light.
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents, light.
Specifications
Specifications
| Melting Point | 80°C to 84°C |
| Boiling Point | 267°C |
| Flash Point | 148°C (298°F) |
| Odor | Pungent |
| Quantity | 25 g |
| Beilstein | 774631 |
| Sensitivity | Light sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 137.18 |
| Percent Purity | ≥97% |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H312-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 202-727-0 |
| RTECSNumber | SL1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction