Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-endo-Hydroxymethylbicyclo[2.2.1]heptyl-2-endo-amine hydrochloride, ≥99%, Thermo Scientific™

Catalog No. AC265911000
Click to view available options
Quantity:
100 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205639-89-0
Molecular Formula C8H16ClNO
Molecular Weight (g/mol) 177.672
MDL Number MFCD00143122
InChI Key JBSSFLCCAAVUFA-ICDZOTBQSA-N
Synonym diendo-3-amino-bicyclo 2.2.1 hept-2-yl-methanol hydrochloride, 3-endo-hydroxymethylbicyclo 2.2.1 heptyl-2-endo-amine hydrochloride, 1r,2r,3s,4s-3-aminobicyclo 2.2.1 heptan-2-yl methanol hydrochloride
PubChem CID 72208316
IUPAC Name [(1R,2R,3S,4S)-3-amino-2-bicyclo[2.2.1]heptanyl]methanol;hydrochloride
SMILES C1CC2CC1C(C2N)CO.Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 177.67
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-endo-Hydroxymethylbicyclo[2.2.1]heptyl-2-endo-amine hydrochloride, 99+%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.