Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxymethylene-2,4-pentanedione, 97+%

Catalog No. AAB2544514
Click to view available options
Quantity:
5 g
25 g

CAS: 33884-41-2 | C8H12O3 | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33884-41-2
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
MDL Number MFCD07368652
InChI Key ITTXGKOHFZJUEX-UHFFFAOYSA-N
PubChem CID 240418
IUPAC Name 3-(ethoxymethylidene)pentane-2,4-dione
SMILES CCOC=C(C(C)=O)C(C)=O

Specifications

Boiling Point 140°C to 142°C (15 mmHg)
Refractive Index 1.484
Quantity 25 g
Synonym 3-ethoxymethylene pentane-2,4-dione, 3-ethoxymethylene-2,4-pentanedione, 3-ethoxymethylidene pentane-2,4-dione, acmc-1cmxl, 3-acetyl-4-ethoxy-3-buten-2-one, 3-ethoxymethylene-pentan-2,4-dione, 2,4-pentanedione,3-ethoxymethylene, 2,4-pentanedione, 3-ethoxymethylene, methyl 1-methylcarbonyl-2-ethoxyvinyl ketone
Formula Weight 156.18
Percent Purity ≥98%
Chemical Name or Material 3-Ethoxymethylene-2,4-pentanedione
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.