Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxypropionitrile 99.0+%, TCI America™
SDP

Catalog No. E029925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E029925ML 25 mL
E0299500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E029925ML Supplier TCI America Supplier No. E029925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2141-62-0
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00001959
InChI Key DCWQZPJHHVLHSV-UHFFFAOYSA-N
Synonym 3-ethoxypropionitrile, propanenitrile, 3-ethoxy, propionitrile, 3-ethoxy, 3-ethoxy-propionitrile, 3-ethoxypropiononitrile, beta-ethoxypropionitrile, 2-ethoxypropionitrile, alpha-ethoxypropionitrile, propionitrile, 2-ethoxy, .beta.-ethoxypropionitrile
PubChem CID 16501
IUPAC Name 3-ethoxypropanenitrile
SMILES CCOCCC#N

Specifications

Color Yellow
Boiling Point 171°C
Quantity 25 mL
Formula Weight 99.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethoxypropionitrile
EINECSNumber (2)-1543
RTECSNumber TZ4680000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.