Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxypropylamine, 99%

Catalog No. AAB2369436
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2369436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2369436 Supplier Thermo Scientific Chemicals Supplier No. B2369436
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6291-85-6 | C5H13NO | 103.165 g/mol

It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

Solubility
Miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6291-85-6
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00008221
InChI Key SOYBEXQHNURCGE-UHFFFAOYSA-N
Synonym 3-ethoxypropylamine, 1-propanamine, 3-ethoxy, 3-ethoxy-1-propanamine, 3-ethoxy-1-propylamine, propylamine, 3-ethoxy, 3-ethoxy-propylamine, 3-aminopropyl ethyl ether, 3-ethyoxypropylamine, 3-ethoxypropyl amine, 3-ethyloxy-propylamine
PubChem CID 22720
IUPAC Name 3-ethoxypropan-1-amine
SMILES CCOCCCN

Specifications

Density 0.86
Boiling Point 136°C to 138°C
Flash Point 32°C (91°F)
Refractive Index 1.4185
Linear Formula CH3CH2O(CH2)3NH2
Quantity 500 g
UN Number UN2733
Beilstein 1732287
Solubility Information Miscible in water.
Formula Weight 103.16
Percent Purity 99%
Chemical Name or Material 3-Ethoxypropylamine
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 228-545-1
RTECSNumber UI2700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.