Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxypropylamine 98.0+%, TCI America™
SDP

Catalog No. E0296500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0296500ML 500 mL
E029625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0296500ML Supplier TCI America Supplier No. E0296500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6291-85-6
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00008221
InChI Key SOYBEXQHNURCGE-UHFFFAOYSA-N
Synonym 3-ethoxypropylamine, 1-propanamine, 3-ethoxy, 3-ethoxy-1-propanamine, 3-ethoxy-1-propylamine, propylamine, 3-ethoxy, 3-ethoxy-propylamine, 3-aminopropyl ethyl ether, 3-ethyoxypropylamine, 3-ethoxypropyl amine, 3-ethyloxy-propylamine
PubChem CID 22720
IUPAC Name 3-ethoxypropan-1-amine
SMILES CCOCCCN

Specifications

Color Colorless
pH 12 (100g/L H2O soln. 20°C)
Boiling Point 132°C
Quantity 500 mL
UN Number 2734
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Ethoxypropylamine
EINECSNumber (2)-0385
RTECSNumber UI2700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.