Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Ethyl-2,4-dimethylpyrrole

Catalog No. D15840225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15840225G 25 g
D1584025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15840225G Supplier Sigma Aldrich Supplier No. D15840225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-ethyl-2,4-dimethylpyrrole can be used as a substrate for the synthesis of:
  • Boradiazaindacene (bodipy dyes) units as a building block for the fabrication of energy transfer cassettes
  • Meso-benzyl and meso-alkyl dipyrrins salts by treating with corresponding acid chloride
  • 2,5-bis[3,5-dimethyl-4-ethylpyrrol-2-yl]cyclohexadiene-1,4-dione by reacting with 1,4-benzoquinone

Specifications

CAS 517-22-6
Density 0.913 g/mL (at 25°C (literature))
Boiling Point 197°C (710 mmHg)
Molecular Formula C8H13N
Refractive Index n20/D 1.496 (literature)
Linear Formula C8H13N
MDL Number MFCD00005222
Quantity 25 g
Synonym 2,4-Dimethyl-3-ethylpyrrole; Kryptopyrrole
Molecular Weight (g/mol) 123.2
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.