Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-5-hexen-3-ol, 97%, Thermo Scientific™

Catalog No. AAB2042214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2042214 25 g
AAB2042222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2042214 Supplier Thermo Scientific Chemicals Supplier No. B2042214

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1907-46-6
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00021834
InChI Key ZNPHYMBSDNDCCM-UHFFFAOYSA-N
Synonym 3-ethyl-5-hexen-3-ol, 5-hexen-3-ol, 3-ethyl, allyl diethyl carbinol, acmc-1buxb, znphymbsdndccm-uhfffaoysa
PubChem CID 137255
IUPAC Name 3-ethylhex-5-en-3-ol
SMILES CCC(O)(CC)CC=C

Specifications

Quantity 25 g
Formula Weight 128.21
Percent Purity 97%
Chemical Name or Material 3-Ethyl-5-hexen-3-ol
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.