Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethynylthiophene, 96%

Catalog No. p-7037222
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55898MD 250 mg
AAH5589803 1 g
AAH5589806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55898MD Supplier Thermo Scientific Chemicals Supplier No. H55898MD
Only null left
Add to Cart

CAS: 67237-53-0 | C6H4S | 108.158 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67237-53-0
Molecular Formula C6H4S
Molecular Weight (g/mol) 108.158
MDL Number MFCD04039973
InChI Key MJHLPKWONJUCFK-UHFFFAOYSA-N
Synonym 3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b
PubChem CID 3548422
IUPAC Name 3-ethynylthiophene
SMILES C#CC1=CSC=C1

Specifications

Density 1.098
Boiling Point 152°C to 153°C
Flash Point 46°C (115°F)
Refractive Index 1.58
Quantity 250 mg
UN Number UN1993
Solubility Information Slightly soluble in water.
Formula Weight 108.16
Percent Purity 96%
Chemical Name or Material 3-Ethynylthiophene
Hazard Category H226-H315-H318-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.