Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-exo-Hydroxymethylbicyclo[2.2.1]heptyl-2-exo-amine, hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 174°C to 175°C |
| Color | Beige |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Quantity | 250 mg |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol |
| Formula Weight | 177.67 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction