Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-2-nitrotoluene, 96%, Thermo Scientific™

Catalog No. p-7055083
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3013-27-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD08458093
InChI Key JSXAJLMUBSEJFF-UHFFFAOYSA-N
Synonym 3-fluoro-2-nitrotoluene, 1-fluoro-3-methyl-2-nitro-benzene, 2-fluoro-6-methylnitrobenzene, benzene, 1-fluoro-3-methyl-2-nitro, fluoronitrotoluene, fluor-nitro-toluole, pubchem18581, 2-nitro-3-fluorotoluene, ksc497k3l, 3-methyl-2-nitrofluorobenzene
PubChem CID 19773803
IUPAC Name 1-fluoro-3-methyl-2-nitrobenzene
SMILES CC1=C(C(=CC=C1)F)[N+](=O)[O-]

Specifications

Melting Point 17°C to 18°C
Color White
Quantity 1 g
UN Number UN2811
Formula Weight 155.13
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material 3-Fluoro-2-nitrotoluene
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.