Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog No. AAH5997003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5997003 1 g
AAH59970MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5997003 Supplier Thermo Scientific Chemicals Supplier No. H5997003
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Boronate esters are air- and chromatography-stable and suitable for spectroscopic study. The Suzuki-Miyaura cross-coupling reaction can be used with boronate esters.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873431-46-0
Molecular Formula C16H23BFNO3
Molecular Weight (g/mol) 307.172
MDL Number MFCD22988989
InChI Key YOJYRVSDQHWBGS-UHFFFAOYSA-N
Synonym 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-fluoro-4-4-morpholinyl benzeneboronic acid pinacol ester
PubChem CID 70975109
IUPAC Name 4-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCOCC3)F

Specifications

Quantity 1 g
Formula Weight 307.17
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.