Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-hydroxy-5-methoxybenzaldehyde, 96%

Catalog No. AAH6647703
Click to view available options
Quantity:
250 mg
1 g

CAS: 79418-78-3 | C8H7FO3 | 170.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79418-78-3
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD02683560
InChI Key OOGOFUKAJDPHDJ-UHFFFAOYSA-N
Synonym 5-fluorovanillin, benzaldehyde, 3-fluoro-4-hydroxy-5-methoxy, 3-fluoro-4-hydroxy-5-methoxy-benzaldehyde, pubchem10464, acmc-209xkq, 3-fluoro-4-hydroxy-5-methoxy benzaldehyde, 3-fluoro-5-methoxy-4-hydroxybenzaldehyde, #, 3-fluoranyl-5-methoxy-4-oxidanyl-benzaldehyde, 3-fluoro-4-hydroxy-5-methyloxy benzaldehyde
PubChem CID 598452
IUPAC Name 3-fluoro-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)F)O

Specifications

Color Brown
Boiling Point 114°C to 118°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 170.14
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 3-Fluoro-4-hydroxy-5-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.