Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-hydroxybenzeneboronic acid, 97%

Catalog No. p-7035393
Click to view available options
Quantity:
250 mg
1 g

CAS: 182344-14-5 | C6H6BFO3 | 155.919 g/mol

3-Fluoro-4-hydroxybenzeneboronic acid is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-4-hydroxybenzeneboronic acid is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182344-14-5
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.919
MDL Number MFCD06659838
InChI Key OYNDLOJPYURCJG-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxyphenyl boronic acid, 4-borono-2-fluorophenol, boronic acid, 3-fluoro-4-hydroxyphenyl, 3-fluoro-4-hydroxybenzeneboronic acid, pubchem18807, acmc-1c2s7, 3-fluoro-4-hydroxy-phenylboronic acid, 4-hydroxy-3-fluorophenyl boronic acid, 3-fluoro-4-hydroxyphenyl-boronic acid
PubChem CID 23005361
IUPAC Name (3-fluoro-4-hydroxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)O)F)(O)O

Specifications

Melting Point 252°C to 256°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 155.92
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-hydroxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.