Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. F05431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05431G 1 g
F05435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05431G Supplier TCI America Supplier No. F05431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 403-20-3
Molecular Formula C8H6FO3
Molecular Weight (g/mol) 169.13
MDL Number MFCD00060675
InChI Key HYNNNQDQEORWEU-UHFFFAOYSA-M
Synonym 3-fluoro-4-methoxy-benzoic acid, 3-fluoro-p-anisic acid, 3-fluoro-4-methoxy benzoic acid, 3-fluoro-4-methoxybenzoicacid, pubchem1393, opera_id_903, acmc-209jd1, ksc494s2f, 3-fluoro-4-methoxy benzoicacid, rarechem al bo 0870
PubChem CID 2733401
IUPAC Name 3-fluoro-4-methoxybenzoate
SMILES COC1=CC=C(C=C1F)C([O-])=O

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 1 g
Formula Weight 170.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-methoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.