Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-hydroxypyridine 98.0+%, TCI America™
SDP

Catalog No. F10201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10201G 1 g
F1020200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10201G Supplier TCI America Supplier No. F10201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 209328-55-2
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.09
MDL Number MFCD03095199
InChI Key IUFLFKASIHPKNZ-UHFFFAOYSA-N
Synonym 3-fluoro-5-hydroxypyridine, 5-fluoro-3-pyridinol, 3-pyridinol, 5-fluoro, 5-fluoro-3-hydroxypyridine, pubchem6625, acmc-209ffy, 5-fluoranylpyridin-3-ol, ksc201g7t, 3-pyridinol, 5-fluoro-9ci
PubChem CID 2762906
IUPAC Name 5-fluoropyridin-3-ol
SMILES OC1=CC(F)=CN=C1

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 1 g
Formula Weight 113.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-5-hydroxypyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.