Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH6284103
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1416367-00-4
Molecular Formula C13H18BFO3
Molecular Weight (g/mol) 252.09
MDL Number MFCD16996298
InChI Key DGHRKRRFVKPHDQ-UHFFFAOYSA-N
Synonym 2-3-fluoro-5-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-5-methoxyphenylboronic acid pinacol ester, 3-fluoro-5-methoxybenzeneboronic acid pinacol ester
PubChem CID 72221149
IUPAC Name 2-(3-fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES COC1=CC(F)=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 252.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-methoxybenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.