Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-N-methylaniline, 97%

Catalog No. AAB2256518
Click to view available options
Quantity:
10 g
50 g

Catalog No. AAB2256518

Supplier: Thermo Scientific Chemicals B2256518

Only null left
Add to Cart

CAS: 1978-37-6 | C7H8FN | 125.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1978-37-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD02683095
InChI Key FHYDHJXZZQCXOX-UHFFFAOYSA-N
Synonym n-methyl-3-fluoroaniline, 3-fluoro-n-methylbenzenamine, 3-fluoro-n-methylanilin, n-methyl-m-fluoroaniline, acmc-209xf6, ksc497i4f, 3-fluoro-phenyl-methyl-amine, 3-fluoro-n-methylaniline, benzenamine, 3-fluoro-n-methyl
PubChem CID 2759011
IUPAC Name 3-fluoro-N-methylaniline
SMILES CNC1=CC(=CC=C1)F

Specifications

Density 1.11
Boiling Point 94°C (19 mmHg)
Flash Point 86°C (187°F)
Odor Characteristic
Quantity 50 g
UN Number UN2810
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-N-methylaniline
Hazard Category H227-H302+H332-H311-H315-H319-H373
Hazard Statement GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.