Alfa Aesar™ 3-Fluorobenzaldehyde, 97%

Manufacturer: Alfa Aesar™ A1139609

Quantity 10g

 View more versions of this product

Catalog No. AAA1139609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 456-48-4
    Molecular Formula C7H5FO
    Formula Weight 124.11
    MDL Number MFCD00003348
    UN Number UN1989
    Beilstein 970178
    Chemical Name or Material 3-Fluorobenzaldehyde

    Specifications

    Formula Weight 124.11
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    UN Number UN1989
    Quantity 10g
    MDL Number MFCD00003348
    Beilstein 970178
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H226-H315-H319-H335
    Toxic if swallowed.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
    Flash Point 56°C (132°F)
    Boiling Point 173°C
    Molecular Formula C7H5FO
    CAS 456-48-4
    Chemical Name or Material 3-Fluorobenzaldehyde
    Assay 97%

    GHS H Statement
    H301-H226-H315-H319-H335
    Toxic if swallowed.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit