Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzaldehyde, 98+%

Catalog No. p-3736517
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle

CAS: 456-48-4 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-48-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00003348
InChI Key PIKNVEVCWAAOMJ-UHFFFAOYSA-N
Synonym m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
PubChem CID 68009
IUPAC Name 3-fluorobenzaldehyde
SMILES C1=CC(=CC(=C1)F)C=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 66°C to 68°C (20.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5170 to 1.519
Linear Formula FC6H4CHO
Quantity 10 mL
Beilstein 07, II, 177
Specific Gravity 1.17
Formula Weight 124.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-266-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.