Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH5574003
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1524-40-9
Molecular Formula C6H6FNO2S
Molecular Weight (g/mol) 175.177
MDL Number MFCD00042284
InChI Key CRINBBOGNYCAOV-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1-sulfonamide, benzenesulfonamide, 3-fluoro, 3-fluorobenzenesulphonamide, 3-fbsa, 2weo, pubchem11762, acmc-20a5kf, 3-fluoranylbenzenesulfonamide, ksc180k2p, 3-fluorobenzenesulfonamide
PubChem CID 150924
IUPAC Name 3-fluorobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)F

Specifications

Melting Point 124°C to 129°C
Color White to Brown
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 175.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluorobenzenesulfonamide
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.