Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Fluorobenzoyl Chloride 97.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
250 g
Specifications
Specifications
| Melting Point | -30°C |
| Boiling Point | 91°C |
| Quantity | 250 g |
| UN Number | 3265 |
| Formula Weight | 158.56 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Fluorobenzoyl Chloride |
Safety and Handling
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction