Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzyl chloride, 97%

Catalog No. p-7020636
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 456-42-8 | C7H6ClF | 144.573 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-42-8
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000904
InChI Key XBDXMDVEZLOGMC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl chloride, 1-chloromethyl-3-fluorobenzene, m-fluorobenzyl chloride, benzene, 1-chloromethyl-3-fluoro, 3-fluorobenzylchloride, alpha-chloro-3-fluorotoluene, alpha-chloro-m-fluorotoluene, fluorobenzyl-3 chloride, toluene, .alpha.-chloro-m-fluoro, .alpha.-chloro-m-fluorotoluene
PubChem CID 9974
IUPAC Name 1-(chloromethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CCl

Specifications

Density 1.213
Boiling Point 176°C to 177°C
Flash Point 58°C (136°F)
Odor Pungent
Refractive Index 1.512
Quantity 5 g
UN Number UN2920
Beilstein 742265
Formula Weight 144.58
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 207-264-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.