Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzylamine 98.0+%, TCI America™
SDP

Catalog No. F053925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F053925G 25 g
F05395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F053925G Supplier TCI America Supplier No. F053925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-82-3
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
InChI Key QVSVMNXRLWSNGS-UHFFFAOYSA-N
Synonym 3-fluorobenzylamine, 3-fluorophenyl methanamine, m-fluorobenzylamine, benzenemethanamine, 3-fluoro, 1-3-fluorophenyl methanamine, 3-fluoro-benzylamine, benzylamine, m-fluoro, 3-fluorophenyl methylamine, m-fluorobenzyl amine, 3-fluorobenzyl amine
PubChem CID 66853
IUPAC Name (3-fluorophenyl)methanamine
SMILES C1=CC(=CC(=C1)F)CN

Specifications

Color Yellow
Boiling Point 82°C
Quantity 25 g
UN Number 2735
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.