Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Fluorophenylboronic acid, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Color | White |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Melting Point | 211.0°C to 212.0°C |
Quantity | 1g |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Linear Formula | FC6H4B(OH)2 |
Solubility Information | Solubility in water: .. Other solubilities: soluble in methanol and thf |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/
GHS Signal Word: Warning