Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenylboronic acid, 97%

Catalog No. AC345640050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 768-35-4 | C6H6BFO2 | 139.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-35-4
Molecular Formula C6H6BFO2
Molecular Weight (g/mol) 139.92
MDL Number MFCD00236042
InChI Key KNXQDJCZSVHEIW-UHFFFAOYSA-N
Synonym 3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid
PubChem CID 2733986
IUPAC Name (3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=CC(F)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 211.0°C to 212.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FC6H4B(OH)2
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol and thf
Formula Weight 139.92
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.