Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoropyridine N-oxide, 98%, Thermo Scientific™

Catalog No. AAH2576506
Click to view available options
Quantity:
5g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoropyridine N-oxide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 695-37-4
Molecular Formula C5H4FNO
Quantity 5g
MDL Number MFCD03095181
Synonym 3-fluoropyridine 1-oxide, 3-fluoropyridine n-oxide, 3-fluoropyridine-n-oxide, pyridine, 3-fluoro-, 1-oxide, 3-fluoropyridin-1-ium-1-olate, 3-fluoropyridinen-oxide, pubchem14114, 1-oxido-3-fluoropyridine, 3-fluoropyridine-1-oxide, 3-fluoro-pyridine 1-oxide
Solubility Information Slightly soluble in water.
InChI Key QVGBDRDOWKIYHK-UHFFFAOYSA-N
SMILES C1=CC(=C[N+](=C1)[O-])F
IUPAC Name 3-fluoro-1-oxidopyridin-1-ium
Molecular Weight (g/mol) 113.091
PubChem CID 123084
Formula Weight 113.09
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine N-oxide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.