Learn More
Labconco™ 3 ft. Logic™ Class II, Type A2 Biosafety Cabinet, US Model
Shop All Labconco Corporation Products

Description
Performance Features:
- Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
- Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
- Full, six-year warranty on all Class II biosafety cabinets
- Optional Configurations
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Telescoping base stand with shelf and leveling feet
Specifications
Specifications
| Description | 3 ft. Logic Class II, Type A2 Biosafety Cabinet |
| Width (Metric) Interior | 92.7 cm |
| Product Type | Class II, Type A2 Bio Safety Cabinet |
| Plug Type | North America (115 V, 15 A) |
| Sash Opening (Metric) | 25 cm |
| Width (English) Exterior | 42.4 in. |
| Width (Metric) Exterior | 107.7 cm |
| Includes | Cabinet, Accessory Package, Base Stand |
| Sash Opening (English) | 10 in. |
| Width (English) Interior | 36.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.