Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 3 ft. Logic™ Class II, Type A2 Biosafety Cabinet, US Model
SDP

Catalog No. 01812660 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Cabinet
Cabinet, Accessory Package
Cabinet, Accessory Package, Base Stand
Cabinet, Base Stand
Plug Type:
North America (115 V, 15 A)
North America (230 V)
Sash Opening (Metric):
20 cm
25 cm
30.5 cm
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Plug Type Sash Opening (Metric)
01-812-660 Cabinet North America (230 V) 25 cm
01-812-645 Cabinet North America (115 V, 15 A) 20 cm
01-812-643 Cabinet, Base Stand North America (115 V, 15 A) 20 cm
01-812-641 Cabinet, Accessory Package North America (115 V, 15 A) 20 cm
01-812-639 Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 20 cm
01-812-659 Cabinet, Base Stand North America (115 V, 15 A) 25 cm
01-812-657 Cabinet, Accessory Package North America (115 V, 15 A) 25 cm
01-812-655 Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 25 cm
01-812-653 Cabinet North America (115 V, 15 A) 30.5 cm
01-812-651 Cabinet, Base Stand North America (115 V, 15 A) 30.5 cm
01-812-649 Cabinet, Accessory Package North America (115 V, 15 A) 30.5 cm
01-812-647 Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 30.5 cm
01-812-644 Cabinet North America (230 V) 20 cm
01-812-642 Cabinet, Base Stand North America (230 V) 20 cm
01-812-640 Cabinet, Accessory Package North America (230 V) 20 cm
01-812-638 Cabinet, Accessory Package, Base Stand North America (230 V) 20 cm
01-812-658 Cabinet, Base Stand North America (230 V) 25 cm
01-812-656 Cabinet, Accessory Package North America (230 V) 25 cm
01-812-654 Cabinet, Accessory Package, Base Stand North America (230 V) 25 cm
01-812-652 Cabinet North America (230 V) 30.5 cm
01-812-650 Cabinet, Base Stand North America (230 V) 30.5 cm
01-812-648 Cabinet, Accessory Package North America (230 V) 30.5 cm
01-812-646 Cabinet, Accessory Package, Base Stand North America (230 V) 30.5 cm
13-620-463 Cabinet North America (115 V, 15 A) 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 01-812-660 Supplier Labconco™ Supplier No. 302310012
Add to Cart
Add to Cart
Request a Quote

Logic Class II, Type A2 Biosafety Cabinets offer personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. All models ensure the highest standard of safety and are listed to NSF/ANSI 49. Logic cabinets maximize safety, performance, and comfort.

Performance Features:

  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
  • Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
  • Full, six-year warranty on all Class II biosafety cabinets
  • Optional Configurations
  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
  • Telescoping base stand with shelf and leveling feet

Specifications

Description 3 ft. Logic Class II, Type A2 Biosafety Cabinet
Width (Metric) Interior 92.7 cm
Product Type Class II, Type A2 Bio Safety Cabinet
Model Logic A2
Plug Type North America (230 V)
Height (English) 61.9 in.
Sash Opening (Metric) 25 cm
Width (English) Exterior 42.4 in.
Width (Metric) Exterior 107.7 cm
Includes Cabinet
Height (Metric) 157.2 cm
Sash Opening (English) 10 in.
Width (English) Interior 36.5 in.
Height (English) Interior 25.7 in.
Height (Metric) Interior 65.3 cm
Depth (English) 32 in.
Depth (Metric) 81.3 cm
Voltage 230 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.