Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco 3 ft. Nexus Horizontal Clean Bench
GSA_VASDP

Catalog No. 17195004 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Epoxy-coated Steel
Stainless Steel
Voltage:
115 V
Includes:
Base Stand
UV Lamp
UV Lamp and Base Stand
Depth (English) Interior:
21 in.
26 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Voltage Includes Depth (English) Interior
17-195-004 Stainless Steel 115 V Base Stand 26 in.
17-194-979 Epoxy-coated Steel 115 V UV Lamp 21 in.
17-195-000 Stainless Steel 115 V UV Lamp 21 in.
17-195-064 Epoxy-coated Steel 115 V Base Stand 21 in.
17-195-043 Stainless Steel 115 V Base Stand 21 in.
17-195-083 Stainless Steel 115 V UV Lamp and Base Stand 21 in.
17-195-050 Stainless Steel 115 V UV Lamp and Base Stand 26 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-195-004 Supplier Labconco Supplier No. 5331600
Add to Cart
Add to Cart
Request a Quote

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.

  • 3 ft. Nexus Horizontal Clean Bench
  • Internal work area
  • Standard interior with Epoxy-coated steel diffuser and non-shedding PVC work surface
  • ISO Class 4 laminar airflow is 10 times cleaner than the industry standard ISO Class 5
  • Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Horizontal Clean Bench
Coating Epoxy
Filter Type HEPA
Lighting Type LED
Material Stainless Steel
Color Optic White
Depth (English) Interior 26 in.
Depth (Metric) Interior 66.6 cm
Height (English) 50.1 in.
Height (English) Interior 30.2 in.
Height (Metric) 127.5 cm
Height (Metric) Interior 76.7 cm
Includes Base Stand
IP Rating None
Plug Type NEMA 5-15P
Voltage 115 V
Width (English) 39.4 in.
Width (English) Interior 37.0 in.
Width (Metric) 100.1 cm
Width (Metric) Interior 93.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.