Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3% Glutaraldehyde in 0.1 Phosphate Buffer, Cenmed™

Click to view available options
Quantity:
500 x 5.2 mL
Description
- Specialized solution is meticulously formulated to achieve optimal results in tissue preservation
- 3% concentration, combined with the phosphate buffer, ensures reliable and effective fixation for various applications
- Packaged in a convenient 5.2 mL vial, this solution offers precise measurements, enhancing ease of use and accuracy in your experiments
Specifications
Specifications
Physical Form | Liquid |
CAS | 111-30-8 |
Color | Clear |
Quantity | 500 x 5.2 mL |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction