Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Heptanone, 98%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
106-35-4
C7H14O
114.19
NGAZZOYFWWSOGK-UHFFFAOYSA-N
3-heptanone,butyl ethyl ketone,n-butyl ethyl ketone,aethylbutylketon,ethylbutylcetone,ethyl n-butyl ketone,ethylbutylketon,etilbutilchetone,eptan-3-one,ethyl butyl ketone
7802
CHEBI:50139
heptan-3-one
CCCCC(=O)CC
106-35-4
C7H14O
114.19
NGAZZOYFWWSOGK-UHFFFAOYSA-N
3-heptanone,butyl ethyl ketone,n-butyl ethyl ketone,aethylbutylketon,ethylbutylcetone,ethyl n-butyl ketone,ethylbutylketon,etilbutilchetone,eptan-3-one,ethyl butyl ketone
7802
CHEBI:50139
heptan-3-one
CCCCC(=O)CC
Packaging Glass bottle
Refractive Index 1.4075 to 1.4095
Infrared Spectrum Authentic
Flash Point 41°C
Quantity 250mL
Melting Point -39.0°C
Color Yellow
Specific Gravity 0.81
Boiling Point 146.0°C to 149.0°C
Density 0.8100g/mL
Assay Percent Range 98%
Linear Formula CH3(CH2)3COC2H5
Beilstein 01,699
Solubility Information Solubility in water: 3.3g/L water (20° c). Other solubilities: soluble in alcohol and ether
Formula Weight 114.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Heptanone
Show More Show Less