Alfa Aesar™ 3-Hydroxy-3-methyl-2-butanone, 90+%

Manufacturer: Alfa Aesar™ 4372422

Quantity 100g

 View more versions of this product

Catalog No. AA4372422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 115-22-0
    Molecular Formula C5H10O2
    Formula Weight 102.13
    MDL Number MFCD00004460
    UN Number UN1224
    Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone

    Specifications

    Formula Weight 102.13
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00004460
    CAS 115-22-0
    UN Number UN1224
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Molecular Formula C5H10O2
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    Density 0.96g/mL
    Flash Point 42°C (107.6°F)
    Boiling Point 140° to 141°C
    Refractive Index 1.4150
    Physical Form Liquid
    Quantity 100g
    Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone
    Assay ≥90%

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit