Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-3-methyl-2-butanone 92%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
115-22-0
C5H10O2
102.13
MFCD00004460
BNDRWEVUODOUDW-UHFFFAOYSA-N
3-hydroxy-3-methyl-2-butanone,3-methylacetoin,methylacetoin,2-butanone, 3-hydroxy-3-methyl,dimethylacetylcarbinol,2-hydroxy-2-methyl-3-butanone,2-methyl-2-hydroxybutan-3-one,1-hydroxy-1-methylethyl methyl ketone,3-hydroxy-3-methyl-butan-2-one,3-hydroxy-3-ethylbutanone
8261
3-hydroxy-3-methylbutan-2-one
CC(=O)C(C)(C)O
115-22-0
C5H10O2
102.13
MFCD00004460
BNDRWEVUODOUDW-UHFFFAOYSA-N
3-hydroxy-3-methyl-2-butanone,3-methylacetoin,methylacetoin,2-butanone, 3-hydroxy-3-methyl,dimethylacetylcarbinol,2-hydroxy-2-methyl-3-butanone,2-methyl-2-hydroxybutan-3-one,1-hydroxy-1-methylethyl methyl ketone,3-hydroxy-3-methyl-butan-2-one,3-hydroxy-3-ethylbutanone
8261
3-hydroxy-3-methylbutan-2-one
CC(=O)C(C)(C)O
Flash Point 42°C
Boiling Point 140°C to 141°C
Specific Gravity 0.97
Density 0.9700g/mL
Refractive Index 1.4145 to 1.4165
Color Colorless to Yellow or Red
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 90.0
CAS Max % 100.0
Assay Percent Range 92%
Linear Formula (CH3)2C(OH)COCH3
Beilstein 01, 832
Formula Weight 102.13
Physical Form Liquid
Percent Purity ≥90%
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone, 92%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-073-1

TSCA : TSCA