Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-methoxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
621-59-0
C8H8O3
152.15
MFCD00003369
JVTZFYYHCGSXJV-UHFFFAOYSA-N
isovanillin,isovanilline,3-hydroxy-p-anisaldehyde,5-formylguaiacol,benzaldehyde, 3-hydroxy-4-methoxy,iso-vanillin,3-hydroxyanisaldehyde,isovanilin,isovanicaline,3-hydroxy-4-methoxy-benzaldehyde
12127
3-hydroxy-4-methoxybenzaldehyde
COC1=C(C=C(C=C1)C=O)O
621-59-0
C8H8O3
152.15
MFCD00003369
JVTZFYYHCGSXJV-UHFFFAOYSA-N
isovanillin,isovanilline,3-hydroxy-p-anisaldehyde,5-formylguaiacol,benzaldehyde, 3-hydroxy-4-methoxy,iso-vanillin,3-hydroxyanisaldehyde,isovanilin,isovanicaline,3-hydroxy-4-methoxy-benzaldehyde
12127
3-hydroxy-4-methoxybenzaldehyde
COC1=C(C=C(C=C1)C=O)O
Boiling Point 179°C (15.0mmHg)
Infrared Spectrum Authentic
Melting Point 112°C to 116°C
Packaging Glass bottle
Color Brown or Gray to White
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H3(OCH3)CHO
Beilstein 08, 254
Formula Weight 152.15
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-methoxybenzaldehyde
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-694-9