Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-nitrobenzoic acid, 98%

Catalog No. AAA1072414
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAA1072414

Supplier: Thermo Scientific Chemicals A1072414

Only null left
Add to Cart

CAS: 619-14-7 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-14-7
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007110
InChI Key XLDLRRGZWIEEHT-UHFFFAOYSA-N
Synonym 3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
PubChem CID 69265
IUPAC Name 3-hydroxy-4-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O

Specifications

Melting Point 232°C to 236°C
Quantity 25 g
Beilstein 2107564
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-580-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.