Alfa Aesar™ 3-Hydroxy-5-methylpyridine, 97%

Manufacturer: Alfa Aesar™ H3265803

Quantity 1g

 View more versions of this product

Catalog No. AAH3265803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 42732-49-0
    Molecular Formula C6H7NO
    Formula Weight 109.13
    MDL Number MFCD00661297
    Synonym 5-Hydroxy-3-picoline; 5-Methyl-3-pyridinol
    Chemical Name or Material 3-Hydroxy-5-methylpyridine

    Specifications

    Formula Weight 109.13
    Synonym 5-Hydroxy-3-picoline; 5-Methyl-3-pyridinol
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 136° to 140°C
    Quantity 1g
    Molecular Formula C6H7NO
    MDL Number MFCD00661297
    CAS 42732-49-0
    Chemical Name or Material 3-Hydroxy-5-methylpyridine
    Assay 97%

    Description

    Applications
    3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

    Solubility
    Slightly soluble in water.

    Notes
    Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit