Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxyazetidine hydrochloride, 97%, Thermo Scientific™

Catalog No. AC360570010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC360570010 1 g
AC360570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360570010 Supplier Thermo Scientific Chemicals Supplier No. 360570010

Chemical Identifiers

CAS 18621-18-6
Molecular Formula C3H7NO·HCl
Molecular Weight (g/mol) 109.56
InChI Key UQUPQEUNHVVNKW-UHFFFAOYSA-N
Synonym 3-hydroxyazetidine hydrochloride, azetidin-3-ol hydrochloride, 3-hydroxyazetidine hcl, azetidin-3-ol hcl, 3-azetidinol hydrochloride, azetidin-3-ol hydrochloride salt, 3-azetidinol, hydrochloride, 3-hydroxy-azetidine hydrochloride, azetidin-3-ol, hydrochloride salt, 3-hydroxy-1-azetidine hydrochloride
PubChem CID 2759290
IUPAC Name azetidin-3-ol;hydrochloride
SMILES C1C(CN1)O.Cl

Specifications

Melting Point 85.0°C to 90.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 109.56
Percent Purity 97%
Chemical Name or Material 3-Hydroxyazetidine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minu

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.