Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxybenzaldehyde 98.5%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
100-83-4
C7H6O2
122.12
MFCD00003368
IAVREABSGIHHMO-UHFFFAOYSA-N
m-formylphenol,m-hydroxybenzaldehyde,3-formylphenol,benzaldehyde, 3-hydroxy,meta-hydroxybenzaldehyde,benzaldehyde, m-hydroxy,3-hydroxy-benzaldehyde,3-hydroxy benzaldehyde,3-hydroxybenzaldehyd,3-hydroxylbenzaldehyde
101
CHEBI:16207
3-hydroxybenzaldehyde
C1=CC(=CC(=C1)O)C=O
100-83-4
C7H6O2
122.12
MFCD00003368
IAVREABSGIHHMO-UHFFFAOYSA-N
m-formylphenol,m-hydroxybenzaldehyde,3-formylphenol,benzaldehyde, 3-hydroxy,meta-hydroxybenzaldehyde,benzaldehyde, m-hydroxy,3-hydroxy-benzaldehyde,3-hydroxy benzaldehyde,3-hydroxybenzaldehyd,3-hydroxylbenzaldehyde
101
CHEBI:16207
3-hydroxybenzaldehyde
C1=CC(=CC(=C1)O)C=O
Color Brown to Yellow
Packaging Plastic bottle
Boiling Point 191°C (50.0mmHg)
Quantity 500g
Infrared Spectrum Authentic
Melting Point 101°C to 105°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98.50%
Linear Formula HOC6H4CHO
Beilstein 08, 58
Solubility Information Solubility in water: soluble in water
Formula Weight 122.12
Physical Form Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material 3-Hydroxybenzaldehyde, 98.5%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-892-9

RTECSNumber : CU6450000

TSCA : TSCA