Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. H0205100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0205100G 100 g
H020525G 25 g
H0205500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0205100G Supplier TCI America Supplier No. H0205100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002506
InChI Key IJFXRHURBJZNAO-UHFFFAOYSA-N
Synonym m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova
PubChem CID 7420
ChEBI CHEBI:30764
IUPAC Name 3-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 203°C
Color White
Quantity 100 g
Formula Weight 138.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzoic Acid
EINECSNumber (3)-1640
RTECSNumber DH1924980
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.