Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxybutyric acid, 95%

Catalog No. p-3778777
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 300-85-6 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-85-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004546
InChI Key WHBMMWSBFZVSSR-UHFFFAOYSA-N
Synonym 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy
PubChem CID 441
ChEBI CHEBI:20067
IUPAC Name 3-hydroxybutanoic acid
SMILES CC(CC(=O)O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 1.1260g/mL
Boiling Point 118.0°C to 120.0°C (2.0 mmHg)
Flash Point 112°C
Assay Percent Range 95&percent;
Packaging Glass bottle
Refractive Index 1.4420 to 1.4440
Linear Formula CH3CH(OH)CH2CO2H
Quantity 5 g
Beilstein 03, 307
Fieser 14,178
Specific Gravity 1.126
Merck Index 15, 4855
Solubility Information Solubility in water: >10% in water. Other solubilities: soluble in alcohol,ether
Formula Weight 104.11
Percent Purity 95%
Grade Technical
Chemical Name or Material 3-Hydroxybutyric acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advic
EINECSNumber 206-099-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.