Learn More
3-Hydroxybutyric acid, tech.
Description
Industrially, 3-hydroxybutanoic acid is used for the manufacture of biodegradable plastics. It is also acts as a solvent in organoleptics, cosmetics. It is also used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by Escherichia coli.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Density | 1.13 g/mL |
Boiling Point | 130°C (12 mmHg) |
Flash Point | >112°C (234°F) |
Refractive Index | 1.4445 |
Quantity | 25 g |
Sensitivity | Hygroscopic |
Merck Index | 14,4816 |
Solubility Information | Soluble in water, alcohol, ether |
Formula Weight | 104.1 |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 206-099-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.