Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 3-hydroxykynurenine Monoclonal Antibody (P3UI)
GREENER_CHOICE

Catalog No. AMPA6616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6616 Supplier Invitrogen™ Supplier No. MA116616
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: 3-OHKYN modified BSA.

3-Hydroxykynurenine (3-HK) is a metabolite in the kynurenine pathway of tryptophan catabolism, primarily involved in various physiological and pathological processes. It is a branch of the kynurenine monooxygenase (KMO) pathway and has been recognized for its dual role in the central nervous system. Structurally, 3-HK is an intermediate metabolite that undergoes autoxidation, which results in a range of products that may contribute to protein modification and cytotoxicity, especially in the context of oxidative stress. Although 3-HK has antioxidant and free radical scavenging properties, it is largely characterized as a toxic metabolite that can induce oxidative damage and cell death. The neuronal damage associated with 3-HK is mediated by free radicals rather than glutamate receptors. Altered levels of 3-HK have been observed in patients with various disorders, suggesting its potential involvement in neurodegenerative diseases. Research continues to explore the balance between its damaging and protective roles, with implications for therapeutic interventions targeting oxidative stress and neuroprotection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 3-hydroxykynurenine
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone P3UI
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene Alias C10H12N2O4
Host Species Mouse
Immunogen 3-OHKYN modified KLH.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.