Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Hydroxymethyl)-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193898 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 3-(Hydroxymethyl)-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 908142-03-0
Color White-Yellow
Molecular Formula C8H11BO4
Quantity 1 g
MDL Number MFCD14155851
Synonym 3-hydroxymethyl-4-methoxyphenyl boronic acid, 3-hydroxymethyl-4-methoxyphenylboronic acid, ksc005q4b, 3-hydroxymethyl-4-methoxybenzeneboronic acid, 3-hydroxymethyl-4-methoxyphenyl boronicacid, b-3-hydroxymethyl-4-methoxyphenyl-boronic acid, boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
InChI Key DOQUCBSZQOPLGT-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)OC)CO)(O)O
IUPAC Name [3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
Molecular Weight (g/mol) 181.982
PubChem CID 53407782
Formula Weight 181.98
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.