Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Hydroxymethyl)benzeneboronic acid pinacol ester

Catalog No. AAH5112606
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 443776-76-9 | C13H19BO3 | 234.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443776-76-9
Molecular Formula C13H19BO3
Molecular Weight (g/mol) 234.102
MDL Number MFCD09266196
InChI Key ZEWWJJQAFTXUIS-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethyl phenylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethylphenylboronic acid pinacol ester, benzenemethanol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb056, 3-hydroxymethyl benzenboronic acid pinacol ester, 3-hydroxymethylphenylboronic acid, pinacol ester, 3-hydroxymethyl benzeneboronic acid pinacol ester
PubChem CID 44118236
IUPAC Name [3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CO

Specifications

Quantity 5 g
Formula Weight 234.1
Chemical Name or Material 3-(Hydroxymethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.