Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxyphenylacetic Acid 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
621-37-4
C8H8O3
152.15
MFCD00004337
FVMDYYGIDFPZAX-UHFFFAOYSA-N
3-hydroxyphenylacetic acid,2-3-hydroxyphenyl acetic acid,3-hydroxyphenyl acetic acid,3-hydroxybenzeneacetic acid,3-hydroxyphenylacetate,m-hydroxyphenylacetic acid,m-hydroxyphenyl acetic acid,benzeneacetic acid, 3-hydroxy,metahydroxy phenylacetic acid,acetic acid, m-hydroxyphenyl
12122
CHEBI:17445
2-(3-hydroxyphenyl)acetic acid
C1=CC(=CC(=C1)O)CC(=O)O
621-37-4
C8H8O3
152.15
MFCD00004337
FVMDYYGIDFPZAX-UHFFFAOYSA-N
3-hydroxyphenylacetic acid,2-3-hydroxyphenyl acetic acid,3-hydroxyphenyl acetic acid,3-hydroxybenzeneacetic acid,3-hydroxyphenylacetate,m-hydroxyphenylacetic acid,m-hydroxyphenyl acetic acid,benzeneacetic acid, 3-hydroxy,metahydroxy phenylacetic acid,acetic acid, m-hydroxyphenyl
12122
CHEBI:17445
2-(3-hydroxyphenyl)acetic acid
C1=CC(=CC(=C1)O)CC(=O)O
Infrared Spectrum Authentic
Color Beige-Brown to White
Melting Point 128.0°C to 131.0°C
Quantity 5g
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula HOC6H4CH2CO2H
Beilstein 10, 189
Solubility Information Solubility in water: soluble in water
Formula Weight 152.15
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material 3-Hydroxyphenylacetic acid, 99+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-684-4

TSCA : TSCA