Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Hydroxypicolinic acid matrix substance for MALDI-MS, ≥99% (HPLC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
CAS | 874-24-8 |
Linear Formula | C6H5 NO3 |
MDL Number | MFCD00006294 |
Melting Point | 213°C to 218°C |
Synonym | 3-HPA; 3-Hydroxypyridine-2-carboxylic acid |
Type | Organic Standard |
Quantity | 250 mg |
Formula Weight | 139.11 |
Percent Purity | ≥99% (HPLC) |
Grade | Matrix Substance for MALDI-MS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction