Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypropanesulfonic Acid (contains varying amounts of 3,3'-Oxydipropanesulfonic Acid), TCI America™
SDP

Catalog No. h059725g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H059725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H059725G Supplier TCI America Supplier No. H059725G
Add to Cart
Add to Cart

(ca. 80% in Water, ca. 7.8mol)

Chemical Identifiers

CAS 15909-83-8
Molecular Formula C3H8O4S
Molecular Weight (g/mol) 140.15
MDL Number MFCD00066499
InChI Key WQPMYSHJKXVTME-UHFFFAOYSA-N
PubChem CID 85181
IUPAC Name 3-hydroxypropane-1-sulfonic acid
SMILES OCCCS(O)(=O)=O

Specifications

Quantity 25 g
UN Number 3265
Formula Weight 140.15
Physical Form Liquid
Chemical Name or Material 3-Hydroxypropanesulfonic Acid (contains varying amounts of 3,3′-Oxydipropanesulfonic Acid) (ca
EINECSNumber (2)-1645
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.